Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 1F7]

Proveedor: OriGene
CF505332
ORIGCF505332EA 786 EUR
ORIGCF505332
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 1F7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CCT8L2
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR