Anti-MVK Mouse Monoclonal Antibody [clone: 1D7]

Proveedor: OriGene
CF502934
ORIGCF502934EA 764 EUR
ORIGCF502934
Anti-MVK Mouse Monoclonal Antibody [clone: 1D7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MVK
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR