Anti-TULP3 Mouse Monoclonal Antibody [clone: 4E5]

Proveedor: OriGene
CF504149
ORIGCF504149EA 786 EUR
ORIGCF504149
Anti-TULP3 Mouse Monoclonal Antibody [clone: 4E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: TULP3
Clonality: Monoclonal
Clone: 4E5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR