Anti-PNPO Mouse Monoclonal Antibody [clone: 1H9]

Proveedor: OriGene
ORIGCF503356EA 764 EUR
ORIGCF503356
Anti-PNPO Mouse Monoclonal Antibody [clone: 1H9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: PNPO
Clonality: Monoclonal
Clone: 1H9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR