Anti-PDE1B Mouse Monoclonal Antibody [clone: 1D6]

Proveedor: OriGene
ORIGCF503336EA 764 EUR
ORIGCF503336
Anti-PDE1B Mouse Monoclonal Antibody [clone: 1D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PDE1B
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR