Anti-NDEL1 Mouse Monoclonal Antibody [clone: 5C2]

Proveedor: OriGene
CF503313
ORIGCF503313EA 764 EUR
ORIGCF503313
Anti-NDEL1 Mouse Monoclonal Antibody [clone: 5C2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: NDEL1
Clonality: Monoclonal
Clone: 5C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR