Anti-PTPN1 Mouse Monoclonal Antibody [clone: 1D10]

Proveedor: OriGene
CF503310
ORIGCF503310EA 786 EUR
ORIGCF503310
Anti-PTPN1 Mouse Monoclonal Antibody [clone: 1D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PTPN1
Clonality: Monoclonal
Clone: 1D10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR