Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 4F4]

Proveedor: OriGene
CF505307
ORIGCF505307EA 786 EUR
ORIGCF505307
Anti-CCT8L2 Mouse Monoclonal Antibody [clone: 4F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CCT8L2
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR