Anti-B3GNT2 Mouse Monoclonal Antibody [clone: 3A9]

Proveedor: OriGene
CF505283
ORIGCF505283EA 764 EUR
ORIGCF505283
Anti-B3GNT2 Mouse Monoclonal Antibody [clone: 3A9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: B3GNT2
Clonality: Monoclonal
Clone: 3A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR