Anti-FATE1 Mouse Monoclonal Antibody [clone: 6G10]

Proveedor: OriGene
ORIGCF505265EA 764 EUR
ORIGCF505265
Anti-FATE1 Mouse Monoclonal Antibody [clone: 6G10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: FATE1
Clonality: Monoclonal
Clone: 6G10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR