Anti-ALDOB Mouse Monoclonal Antibody [clone: 3E10]

Proveedor: OriGene
CF502909
ORIGCF502909EA 764 EUR
ORIGCF502909
Anti-ALDOB Mouse Monoclonal Antibody [clone: 3E10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDOB
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR