Anti-CST4 Mouse Monoclonal Antibody [clone: 2C2]

Proveedor: OriGene
CF504095
ORIGCF504095EA 786 EUR
ORIGCF504095
Anti-CST4 Mouse Monoclonal Antibody [clone: 2C2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CST4
Clonality: Monoclonal
Clone: 2C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR