Anti-GSTO2 Mouse Monoclonal Antibody [clone: 4H1]

Proveedor: OriGene
CF504085
ORIGCF504085EA 764 EUR
ORIGCF504085
Anti-GSTO2 Mouse Monoclonal Antibody [clone: 4H1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GSTO2
Clonality: Monoclonal
Clone: 4H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR