Anti-NMT2 Mouse Monoclonal Antibody [clone: 2E1]

Proveedor: OriGene
CF504072
ORIGCF504072EA 786 EUR
ORIGCF504072
Anti-NMT2 Mouse Monoclonal Antibody [clone: 2E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: NMT2
Clonality: Monoclonal
Clone: 2E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR