Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 1F4]

Proveedor: OriGene
CF506015
ORIGCF506015EA 764 EUR
ORIGCF506015
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 1F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR