Anti-PARVA Mouse Monoclonal Antibody [clone: 1H2]

Proveedor: OriGene
ORIGCF505991EA 764 EUR
ORIGCF505991
Anti-PARVA Mouse Monoclonal Antibody [clone: 1H2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PARVA
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR