Anti-TRIM38 Mouse Monoclonal Antibody [clone: 14B4]

Proveedor: OriGene
CF505983
ORIGCF505983EA 764 EUR
ORIGCF505983
Anti-TRIM38 Mouse Monoclonal Antibody [clone: 14B4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150

Type: Primary
Antigen: TRIM38
Clonality: Monoclonal
Clone: 14B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR