Anti-TRIM38 Mouse Monoclonal Antibody [clone: 16D9]

Proveedor: OriGene
CF505982
ORIGCF505982EA 764 EUR
ORIGCF505982
Anti-TRIM38 Mouse Monoclonal Antibody [clone: 16D9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150

Type: Primary
Antigen: TRIM38
Clonality: Monoclonal
Clone: 16D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR