Anti-STK38L Mouse Monoclonal Antibody [clone: 3E10]

Proveedor: OriGene
CF505210
ORIGCF505210EA 786 EUR
ORIGCF505210
Anti-STK38L Mouse Monoclonal Antibody [clone: 3E10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: STK38L
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR