Anti-SENP2 Mouse Monoclonal Antibody [clone: 1H2]

Proveedor: OriGene
CF504817
ORIGCF504817EA 764 EUR
ORIGCF504817
Anti-SENP2 Mouse Monoclonal Antibody [clone: 1H2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SENP2
Clonality: Monoclonal
Clone: 1H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR