Anti-TPSG1 Mouse Monoclonal Antibody [clone: 2H5]

Proveedor: OriGene
CF504026
ORIGCF504026EA 764 EUR
ORIGCF504026
Anti-TPSG1 Mouse Monoclonal Antibody [clone: 2H5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: TPSG1
Clonality: Monoclonal
Clone: 2H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR