Anti-DGKA Mouse Monoclonal Antibody [clone: 3G7]

Proveedor: OriGene
CF504024
ORIGCF504024EA 764 EUR
ORIGCF504024
Anti-DGKA Mouse Monoclonal Antibody [clone: 3G7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DGKA
Clonality: Monoclonal
Clone: 3G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR