Anti-FBXO31 Mouse Monoclonal Antibody [clone: 1A11]

Proveedor: OriGene
CF504012
ORIGCF504012EA 764 EUR
ORIGCF504012
Anti-FBXO31 Mouse Monoclonal Antibody [clone: 1A11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FBXO31
Clonality: Monoclonal
Clone: 1A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR