Anti-TRIM45 Mouse Monoclonal Antibody [clone: 4F1]

Proveedor: OriGene
CF505922
ORIGCF505922EA 786 EUR
ORIGCF505922
Anti-TRIM45 Mouse Monoclonal Antibody [clone: 4F1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200; Immunochemistry: 1:150

Type: Primary
Antigen: TRIM45
Clonality: Monoclonal
Clone: 4F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR