Anti-RNF39 Mouse Monoclonal Antibody [clone: 5F5]

Proveedor: OriGene
ORIGCF505915EA 764 EUR
ORIGCF505915
Anti-RNF39 Mouse Monoclonal Antibody [clone: 5F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RNF39
Clonality: Monoclonal
Clone: 5F5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR