Anti-RNF39 Mouse Monoclonal Antibody [clone: 4D3]

Proveedor: OriGene
ORIGCF505914EA 764 EUR
ORIGCF505914
Anti-RNF39 Mouse Monoclonal Antibody [clone: 4D3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RNF39
Clonality: Monoclonal
Clone: 4D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR