Anti-MAPK9 Mouse Monoclonal Antibody [clone: 4D1]

Proveedor: OriGene
CF505910
ORIGCF505910EA 764 EUR
ORIGCF505910
Anti-MAPK9 Mouse Monoclonal Antibody [clone: 4D1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK9
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR