Anti-OTUB1 Mouse Monoclonal Antibody [clone: 2H1]

Proveedor: OriGene
CF505158
ORIGCF505158EA 764 EUR
ORIGCF505158
Anti-OTUB1 Mouse Monoclonal Antibody [clone: 2H1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: OTUB1
Clonality: Monoclonal
Clone: 2H1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR