Anti-SDCBP Mouse Monoclonal Antibody [clone: 2H6]

Proveedor: OriGene
CF504796
ORIGCF504796EA 764 EUR
ORIGCF504796
Anti-SDCBP Mouse Monoclonal Antibody [clone: 2H6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: SDCBP
Clonality: Monoclonal
Clone: 2H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR