Anti-SPINT1 Mouse Monoclonal Antibody [clone: 1E2]

Proveedor: OriGene
CF504793
ORIGCF504793EA 764 EUR
ORIGCF504793
Anti-SPINT1 Mouse Monoclonal Antibody [clone: 1E2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SPINT1
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR