Anti-SDS Mouse Monoclonal Antibody [clone: 4C3]

Proveedor: OriGene
ORIGCF503997EA 786 EUR
ORIGCF503997
Anti-SDS Mouse Monoclonal Antibody [clone: 4C3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: SDS
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR