Anti-PFKP Mouse Monoclonal Antibody [clone: 2E8]

Proveedor: OriGene
CF503984
ORIGCF503984EA 764 EUR
ORIGCF503984
Anti-PFKP Mouse Monoclonal Antibody [clone: 2E8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PFKP
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR