Anti-SESTD1 Mouse Monoclonal Antibody [clone: 3A11]

Proveedor: OriGene
CF505833
ORIGCF505833EA 786 EUR
ORIGCF505833
Anti-SESTD1 Mouse Monoclonal Antibody [clone: 3A11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: SESTD1
Clonality: Monoclonal
Clone: 3A11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR