Anti-RSAD2 Mouse Monoclonal Antibody [clone: 4D12]

Proveedor: OriGene
CF505799
ORIGCF505799EA 764 EUR
ORIGCF505799
Anti-RSAD2 Mouse Monoclonal Antibody [clone: 4D12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: RSAD2
Clonality: Monoclonal
Clone: 4D12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR