Anti-BBOX1 Mouse Monoclonal Antibody [clone: 2G1]

Proveedor: OriGene
ORIGCF505788EA 764 EUR
ORIGCF505788
Anti-BBOX1 Mouse Monoclonal Antibody [clone: 2G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: BBOX1
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR