Anti-SLA2 Mouse Monoclonal Antibody [clone: 2F11]

Proveedor: OriGene
ORIGCF505128EA 764 EUR
ORIGCF505128
Anti-SLA2 Mouse Monoclonal Antibody [clone: 2F11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SLA2
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR