Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5H4]

Proveedor: OriGene
CF505959
ORIGCF505959EA 764 EUR
ORIGCF505959
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 5H4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 5H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR