Anti-CTAG1B Mouse Monoclonal Antibody [clone: 4G7]

Proveedor: OriGene
CF505946
ORIGCF505946EA 764 EUR
ORIGCF505946
Anti-CTAG1B Mouse Monoclonal Antibody [clone: 4G7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: CTAG1B
Clonality: Monoclonal
Clone: 4G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR