Anti-ZBED1 Mouse Monoclonal Antibody [clone: 1D5]

Proveedor: OriGene
CF505069
ORIGCF505069EA 764 EUR
ORIGCF505069
Anti-ZBED1 Mouse Monoclonal Antibody [clone: 1D5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ZBED1
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR