Anti-ZNRD1 Mouse Monoclonal Antibody [clone: 1G2]

Proveedor: OriGene
CF505067
ORIGCF505067EA 764 EUR
ORIGCF505067
Anti-ZNRD1 Mouse Monoclonal Antibody [clone: 1G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: ZNRD1
Clonality: Monoclonal
Clone: 1G2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR