Anti-SCFD1 Mouse Monoclonal Antibody [clone: 2E7]

Proveedor: OriGene
CF504662
ORIGCF504662EA 764 EUR
ORIGCF504662
Anti-SCFD1 Mouse Monoclonal Antibody [clone: 2E7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: SCFD1
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR