Anti-RYBP Mouse Monoclonal Antibody [clone: 2B4]

Proveedor: OriGene
ORIGCF504651EA 764 EUR
ORIGCF504651
Anti-RYBP Mouse Monoclonal Antibody [clone: 2B4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RYBP
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR