Anti-HMOX2 Mouse Monoclonal Antibody [clone: 4H8]

Proveedor: OriGene
CF503925
ORIGCF503925EA 786 EUR
ORIGCF503925
Anti-HMOX2 Mouse Monoclonal Antibody [clone: 4H8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HMOX2
Clonality: Monoclonal
Clone: 4H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR