Anti-TAL1 Mouse Monoclonal Antibody [clone: 6H6]

Proveedor: OriGene
CF503919
ORIGCF503919EA 764 EUR
ORIGCF503919
Anti-TAL1 Mouse Monoclonal Antibody [clone: 6H6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TAL1
Clonality: Monoclonal
Clone: 6H6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR