Anti-PDIA4 Mouse Monoclonal Antibody [clone: 6B6]

Proveedor: OriGene
CF503888
ORIGCF503888EA 764 EUR
ORIGCF503888
Anti-PDIA4 Mouse Monoclonal Antibody [clone: 6B6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PDIA4
Clonality: Monoclonal
Clone: 6B6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR