Anti-ERBB2 Mouse Monoclonal Antibody [clone: G9]

Proveedor: OriGene
CF505859
ORIGCF505859EA 764 EUR
ORIGCF505859
Anti-ERBB2 Mouse Monoclonal Antibody [clone: G9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR