Anti-SYT4 Mouse Monoclonal Antibody [clone: 2D3]

Proveedor: OriGene
CF505849
ORIGCF505849EA 786 EUR
ORIGCF505849
Anti-SYT4 Mouse Monoclonal Antibody [clone: 2D3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: SYT4
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR