Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 5C11]

Proveedor: OriGene
CF505838
ORIGCF505838EA 764 EUR
ORIGCF505838
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 5C11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 5C11
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR