Anti-PSMB7 Mouse Monoclonal Antibody [clone: 4E3]

Proveedor: OriGene
ORIGCF504638EA 786 EUR
ORIGCF504638
Anti-PSMB7 Mouse Monoclonal Antibody [clone: 4E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PSMB7
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR