Anti-C6orf64 Mouse Monoclonal Antibody [clone: 2H3]

Proveedor: OriGene
CF504635
ORIGCF504635EA 786 EUR
ORIGCF504635
Anti-C6orf64 Mouse Monoclonal Antibody [clone: 2H3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: C6orf64
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR